The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017433.4 | NP_059129.3 | 20 - 21 | 2624 | 166 |
XM_011519498.2 | XP_011517800.1 | 20 - 21 | 2468 | 166 | |
XM_011519499.1 | XP_011517801.1 | 20 - 21 | 2445 | 166 | |
XM_011519500.2 | XP_011517802.1 | 21 - 22 | 2514 | 166 | |
XM_011519502.1 | XP_011517804.1 | 20 - 21 | 2468 | 166 | |
XM_011519503.1 | XP_011517805.1 | 20 - 21 | 2468 | 166 | |
XM_011519504.1 | XP_011517806.1 | 20 - 21 | 2468 | 166 | |
XM_011519505.1 | XP_011517807.1 | 20 - 21 | 2468 | 166 | |
XM_011519506.2 | XP_011517808.1 | 20 - 21 | 2468 | 166 | |
XM_011519508.2 | XP_011517810.1 | 20 - 21 | 2468 | 166 | |
XM_011519510.1 | XP_011517812.1 | 20 - 21 | 2468 | 166 | |
XM_011519511.2 | XP_011517813.1 | 20 - 21 | 2468 | 166 | |
XM_011519512.1 | XP_011517814.1 | 3 - 4 | 472 | 166 | |
GenBank mRNA | AF229172.1 | - | 20 - 21 | 2442 | 166 |
AK310494.1 | - | 20 - 21 | 2468 | 166 | |
AY101367.1 | - | 20 - 21 | 2442 | 166 | |
BC125272.1 | - | 18 - 19 | 2197 | 166 |