The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012341.2 | NP_036473.2 | 4 - 5 | 530 | 65 |
GenBank mRNA | AF120334.1 | - | 4 - 5 | 482 | 65 |
AF325353.1 | - | 4 - 5 | 459 | 65 | |
AK001548.1 | - | 4 - 5 | 477 | 65 | |
AK001552.1 | - | 4 - 5 | 532 | 65 | |
AK222861.1 | - | 4 - 5 | 477 | 65 | |
AK295231.1 | - | 3 - 4 | 373 | 65 | |
AK301721.1 | - | 4 - 5 | 645 | 65 | |
AK302219.1 | - | 3 - 4 | 335 | 65 | |
BC033784.1 | - | 4 - 5 | 453 | 65 | |
BC038975.2 | - | 4 - 5 | 491 | 65 |