Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO2
Entrez Gene ID
26232
Gene Name
F-box protein 2
Gene Aliases
FBG1, FBX2, Fbs1, NFB42, OCP1
Chromosome Location
Chr.1: 11648361 - 11654831 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012168.5 NP_036300.2 1 - 2 514 163
GenBank mRNA AF187318.1 - 1 - 2 365 160
BC025233.1 - 1 - 2 98 163
BC096747.1 - 1 - 2 114 163

View Full Product Details