Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO2
Entrez Gene ID
26232
Gene Name
F-box protein 2
Gene Aliases
FBG1, FBX2, Fbs1, NFB42, OCP1
Chromosome Location
Chr.1: 11648361 - 11654831 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012168.5 NP_036300.2 3 - 4 1015 78
GenBank mRNA AF174594.1 - 2 - 3 407 78
AF187318.1 - 3 - 4 863 78
AK308531.1 - 4 - 5 997 78
AK313019.1 - 3 - 4 615 78
BC025233.1 - 3 - 4 599 78
BC096747.1 - 3 - 4 615 78

View Full Product Details