Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IPO13
Entrez Gene ID
9670
Gene Name
importin 13
Gene Aliases
IMP13, KAP13, LGL2, RANBP13
Chromosome Location
Chr.1: 43946806 - 43968022 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014652.3 NP_055467.3 15 - 16 3192 104
GenBank mRNA AB018267.1 - 15 - 16 3059 104
AF267987.1 - 15 - 16 2397 104
AK023441.1 - 9 - 10 915 104
AK290173.1 - 15 - 16 2820 104
AL117501.1 - 14 - 15 2189 104
BC008194.1 - 15 - 16 3027 104

View Full Product Details