Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IPO13
Entrez Gene ID
9670
Gene Name
importin 13
Gene Aliases
IMP13, KAP13, LGL2, RANBP13
Chromosome Location
Chr.1: 43946806 - 43968022 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014652.3 NP_055467.3 1 - 2 885 87
GenBank mRNA AB018267.1 - 1 - 2 752 87
AF267987.1 - 1 - 2 90 87
AK290173.1 - 1 - 2 513 87
BC002608.2 - 1 - 2 520 87
BC008194.1 - 1 - 2 720 87

View Full Product Details