Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IPO13
Entrez Gene ID
9670
Gene Name
importin 13
Gene Aliases
IMP13, KAP13, LGL2, RANBP13
Chromosome Location
Chr.1: 43946806 - 43968022 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014652.3 NP_055467.3 3 - 4 1754 55
GenBank mRNA AB018267.1 - 3 - 4 1621 55
AF267987.1 - 3 - 4 959 55
AK290173.1 - 3 - 4 1382 55
AL117501.1 - 2 - 3 751 55
BC002608.2 - 3 - 4 1389 55
BC008194.1 - 3 - 4 1589 55

View Full Product Details