The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319165.1 | NP_001306094.1 | 3 - 4 | 671 | 69 |
NM_001319167.1 | NP_001306096.1 | 3 - 4 | 738 | 69 | |
NM_001319168.1 | NP_001306097.1 | 3 - 4 | 629 | 69 | |
NM_001319170.1 | NP_001306099.1 | 3 - 4 | 741 | 69 | |
NM_001384.4 | NP_001375.2 | 3 - 4 | 681 | 69 | |
XM_017000502.1 | XP_016855991.1 | 3 - 4 | 683 | 69 | |
XM_017000503.1 | XP_016855992.1 | 3 - 4 | 617 | 69 | |
XM_017000506.1 | XP_016855995.1 | 3 - 4 | 725 | 69 | |
GenBank mRNA | AK091303.1 | - | 3 - 4 | 600 | 69 |
AK315506.1 | - | 3 - 4 | 665 | 69 | |
BC001389.2 | - | 3 - 4 | 640 | 69 | |
BC003181.1 | - | 3 - 4 | 658 | 69 | |
BC016956.1 | - | 3 - 4 | 640 | 69 | |
BQ056213.1 | - | 3 - 4 | 655 | 69 | |
BQ215136.1 | - | 3 - 4 | 689 | 69 | |
BT007431.1 | - | 3 - 4 | 486 | 69 | |
CX164803.1 | - | 3 - 4 | 608 | 69 |