The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319165.1 | NP_001306094.1 | 4 - 5 | 1353 | 56 |
NM_001319166.1 | NP_001306095.1 | 4 - 5 | 1090 | 56 | |
NM_001319167.1 | NP_001306096.1 | 4 - 5 | 1420 | 56 | |
NM_001319168.1 | NP_001306097.1 | 4 - 5 | 1311 | 56 | |
NM_001319169.1 | NP_001306098.1 | 3 - 4 | 1139 | 56 | |
NM_001319170.1 | NP_001306099.1 | 4 - 5 | 1423 | 56 | |
NM_001319171.1 | NP_001306100.1 | 4 - 5 | 1394 | 56 | |
NM_001384.4 | NP_001375.2 | 4 - 5 | 1363 | 56 | |
XM_017000502.1 | XP_016855991.1 | 4 - 5 | 1365 | 56 | |
XM_017000503.1 | XP_016855992.1 | 4 - 5 | 1299 | 56 | |
XM_017000504.1 | XP_016855993.1 | 3 - 4 | 1604 | 56 | |
XM_017000505.1 | XP_016855994.1 | 3 - 4 | 1536 | 56 | |
XM_017000506.1 | XP_016855995.1 | 4 - 5 | 1407 | 56 | |
GenBank mRNA | AF053003.1 | - | 4 - 5 | 1329 | 56 |
AK091303.1 | - | 4 - 5 | 1282 | 56 | |
AK297933.1 | - | 3 - 4 | 1112 | 56 | |
AK315506.1 | - | 4 - 5 | 1347 | 56 | |
BC001389.2 | - | 4 - 5 | 1322 | 56 | |
BC003181.1 | - | 4 - 5 | 1340 | 56 | |
BC016956.1 | - | 4 - 5 | 1322 | 56 | |
BT007431.1 | - | 4 - 5 | 1168 | 56 |