The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034023.1 | NP_001029195.1 | 1 - 2 | 74 | 84 |
XM_011541785.2 | XP_011540087.1 | 1 - 2 | 99 | 84 | |
GenBank mRNA | AK289366.1 | - | 1 - 2 | 57 | 84 |
AK293598.1 | - | 1 - 2 | 65 | 84 | |
AK300442.1 | - | 1 - 2 | 62 | 84 | |
BM827551.1 | - | 1 - 2 | 86 | 84 | |
CA842195.1 | - | 1 - 2 | 66 | 84 |