Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIST1H3H
Entrez Gene ID
8357
Gene Name
histone cluster 1, H3h
Gene Aliases
H3/k, H3F1K, H3FK
Chromosome Location
Chr.6: 27810064 - 27810536 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC007518.2 - 1 - 1 1243 144

View Full Product Details