Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM55A
Entrez Gene ID
55529
Gene Name
transmembrane protein 55A
Gene Aliases
-
Chromosome Location
Chr.8: 90994271 - 91040975 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018710.2 NP_061180.1 3 - 4 594 66
XM_011517152.2 XP_011515454.1 3 - 4 594 66
GenBank mRNA AK313783.1 - 3 - 4 507 66
AX775961.1 - 3 - 4 444 66
BC033892.1 - 3 - 4 444 66
CR749733.1 - 2 - 3 158 66

View Full Product Details