The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033341.4 | NP_203127.3 | 1 - 1 | 1939 | 92 |
GenBank mRNA | AF420440.1 | - | 1 - 1 | 1938 | 92 |
BC039318.1 | - | 1 - 1 | 1709 | 92 | |
BC046168.1 | - | 1 - 1 | 1683 | 92 | |
BC071665.1 | - | 1 - 1 | 1487 | 92 |