Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIRC2
Entrez Gene ID
84925
Gene Name
disrupted in renal carcinoma 2
Gene Aliases
RCC4
Chromosome Location
Chr.3: 122795054 - 122892157 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032839.2 NP_116228.1 2 - 3 577 90
GenBank mRNA AK027690.1 - 2 - 3 571 90
AK075158.1 - 2 - 3 494 90
AK291176.1 - 2 - 3 561 90
BC022204.1 - 2 - 3 835 90
BC039821.1 - 2 - 3 548 90

View Full Product Details