Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIRC2
Entrez Gene ID
84925
Gene Name
disrupted in renal carcinoma 2
Gene Aliases
RCC4
Chromosome Location
Chr.3: 122795054 - 122892157 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032839.2 NP_116228.1 5 - 6 1079 99
GenBank mRNA AK027690.1 - 5 - 6 1073 99
AK075158.1 - 5 - 6 996 99
AK291176.1 - 5 - 6 1063 99
BC022204.1 - 5 - 6 1337 99
BC033738.1 - 4 - 5 955 99
BC039821.1 - 5 - 6 1050 99

View Full Product Details