Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIRC2
Entrez Gene ID
84925
Gene Name
disrupted in renal carcinoma 2
Gene Aliases
RCC4
Chromosome Location
Chr.3: 122795054 - 122892157 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032839.2 NP_116228.1 6 - 7 1140 96
GenBank mRNA AK027690.1 - 6 - 7 1134 96
AK075158.1 - 6 - 7 1057 96
AK291176.1 - 6 - 7 1124 96
BC022204.1 - 6 - 7 1398 96
BC033738.1 - 5 - 6 1016 96
BC039821.1 - 6 - 7 1111 96

View Full Product Details