The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004441.4 | NP_004432.1 | 1 - 2 | 428 | 87 |
XM_017005866.1 | XP_016861355.1 | 1 - 2 | 428 | 87 | |
GenBank mRNA | AK291208.1 | - | 1 - 2 | 428 | 87 |
AK309741.1 | - | 1 - 2 | 425 | 87 | |
BC111744.1 | - | 1 - 2 | 224 | 87 | |
DA107464.1 | - | 1 - 2 | 427 | 87 | |
L40636.1 | - | 1 - 2 | 273 | 87 |