Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
Chromosome Location
Chr.15: 49326997 - 49620931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152647.2 NP_689860.2 12 - 13 1405 83
XM_005254213.3 XP_005254270.1 12 - 13 1482 83
XM_005254214.3 XP_005254271.1 12 - 13 1415 83
XM_005254215.3 XP_005254272.1 11 - 12 1446 83
XM_005254216.3 XP_005254273.1 11 - 12 1380 83
XM_006720423.2 XP_006720486.1 13 - 14 1447 83
XM_006720424.2 XP_006720487.1 12 - 13 1482 83
XM_006720426.2 XP_006720489.1 10 - 11 1341 83
XM_006720427.3 XP_006720490.1 13 - 14 1453 83
XM_011521319.2 XP_011519621.1 12 - 13 3365 83
XM_011521320.1 XP_011519622.1 10 - 11 857 83
XM_011521321.2 XP_011519623.1 12 - 13 1482 83
XM_011521322.1 XP_011519624.1 9 - 10 821 83
XM_017021990.1 XP_016877479.1 9 - 10 1239 83
XM_017021991.1 XP_016877480.1 13 - 14 1522 83
XM_017021992.1 XP_016877481.1 10 - 11 1170 83
XM_017021993.1 XP_016877482.1 9 - 10 1239 83
XM_017021994.1 XP_016877483.1 8 - 9 1140 83
XM_017021995.1 XP_016877484.1 12 - 13 1482 83
XM_017021996.1 XP_016877485.1 11 - 12 930 83
GenBank mRNA AK057362.1 - 12 - 13 1403 83

View Full Product Details