Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
Chromosome Location
Chr.15: 49326997 - 49620931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152647.2 NP_689860.2 7 - 8 839 112
XM_005254213.3 XP_005254270.1 7 - 8 916 112
XM_005254214.3 XP_005254271.1 7 - 8 849 112
XM_005254215.3 XP_005254272.1 6 - 7 880 112
XM_005254216.3 XP_005254273.1 7 - 8 916 112
XM_005254219.3 XP_005254276.1 7 - 8 916 112
XM_005254220.3 XP_005254277.1 7 - 8 916 112
XM_006720423.2 XP_006720486.1 8 - 9 881 112
XM_006720424.2 XP_006720487.1 7 - 8 916 112
XM_006720427.3 XP_006720490.1 8 - 9 887 112
XM_011521319.2 XP_011519621.1 7 - 8 2799 112
XM_011521320.1 XP_011519622.1 5 - 6 291 112
XM_011521321.2 XP_011519623.1 7 - 8 916 112
XM_011521322.1 XP_011519624.1 4 - 5 255 112
XM_011521324.1 XP_011519626.1 7 - 8 916 112
XM_011521325.2 XP_011519627.1 7 - 8 916 112
XM_011521326.1 XP_011519628.1 7 - 8 916 112
XM_017021991.1 XP_016877480.1 8 - 9 956 112
XM_017021992.1 XP_016877481.1 5 - 6 604 112
XM_017021995.1 XP_016877484.1 7 - 8 916 112
XM_017021996.1 XP_016877485.1 6 - 7 364 112
GenBank mRNA AK057362.1 - 7 - 8 837 112
BC048125.1 - 7 - 8 841 112
BC048139.1 - 7 - 8 841 112

View Full Product Details