The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152589.1 | NP_689802.1 | 6 - 7 | 642 | 119 |
XM_011537985.1 | XP_011536287.1 | 6 - 7 | 885 | 119 | |
XM_017018886.1 | XP_016874375.1 | 5 - 6 | 582 | 119 | |
XM_017018887.1 | XP_016874376.1 | 6 - 7 | 885 | 119 | |
XM_017018888.1 | XP_016874377.1 | 5 - 6 | 582 | 119 | |
GenBank mRNA | AK093140.1 | - | 6 - 7 | 642 | 119 |
AL833338.1 | - | 6 - 7 | 885 | 119 | |
BC038413.2 | - | 5 - 6 | 583 | 119 |