The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011537985.1 | XP_011536287.1 | 8 - 9 | 1117 | 117 |
XM_017018888.1 | XP_016874377.1 | 7 - 8 | 814 | 117 | |
GenBank mRNA | AL833338.1 | - | 8 - 9 | 1117 | 117 |