Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFYVE21
Entrez Gene ID
79038
Gene Name
zinc finger FYVE-type containing 21
Gene Aliases
HCVP7TP1, ZF21
Chromosome Location
Chr.14: 103715744 - 103733668 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198953.1 NP_001185882.1 1 - 2 239 133
NM_024071.3 NP_076976.1 1 - 2 239 133
GenBank mRNA AK057816.1 - 1 - 2 160 133
AK290519.1 - 1 - 2 152 133
AY395756.1 - 1 - 2 154 133
BC001130.2 - 1 - 2 160 133
BC005999.1 - 1 - 2 175 133
BX248261.1 - 1 - 2 145 133
CX786145.1 - 1 - 2 239 133

View Full Product Details