The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198953.1 | NP_001185882.1 | 7 - 8 | 820 | 58 |
NM_024071.3 | NP_076976.1 | 6 - 7 | 766 | 58 | |
GenBank mRNA | AK001921.1 | - | 1 - 2 | 1331 | 58 |
AK057816.1 | - | 7 - 8 | 741 | 58 | |
AK290519.1 | - | 6 - 7 | 679 | 58 | |
AY395756.1 | - | 6 - 7 | 681 | 58 | |
BC001130.2 | - | 6 - 7 | 687 | 58 | |
BC005999.1 | - | 6 - 7 | 702 | 58 | |
BX248261.1 | - | 6 - 7 | 672 | 58 | |
CX786145.1 | - | 7 - 8 | 820 | 58 |