The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138326.2 | NP_612199.2 | 3 - 4 | 267 | 102 |
XM_005263589.4 | XP_005263646.1 | 4 - 5 | 675 | 102 | |
GenBank mRNA | AB071418.1 | - | 3 - 4 | 200 | 102 |
AK125008.1 | - | 3 - 4 | 267 | 102 | |
BC107420.1 | - | 3 - 4 | 207 | 102 |