The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307983.1 | NP_001294912.1 | 7 - 8 | 801 | 132 |
NM_138326.2 | NP_612199.2 | 7 - 8 | 744 | 132 | |
XM_005263586.4 | XP_005263643.1 | 5 - 6 | 894 | 132 | |
XM_005263588.4 | XP_005263645.1 | 6 - 7 | 616 | 132 | |
XM_005263589.4 | XP_005263646.1 | 8 - 9 | 1152 | 132 | |
XM_005263590.4 | XP_005263647.1 | 4 - 5 | 1458 | 132 | |
XM_011510592.2 | XP_011508894.1 | 7 - 8 | 1102 | 132 | |
XM_017003325.1 | XP_016858814.1 | 7 - 8 | 1099 | 132 | |
XM_017003326.1 | XP_016858815.1 | 7 - 8 | 1749 | 132 | |
GenBank mRNA | AB071418.1 | - | 7 - 8 | 677 | 132 |
AK125008.1 | - | 7 - 8 | 744 | 132 | |
BC016018.1 | - | 7 - 8 | 803 | 132 | |
BC107420.1 | - | 7 - 8 | 684 | 132 | |
KU178859.1 | - | 4 - 5 | 503 | 132 |