Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IARS2
Entrez Gene ID
55699
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
Gene Aliases
CAGSSS, ILERS
Chromosome Location
Chr.1: 220094113 - 220148041 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018060.3 NP_060530.3 15 - 16 2040 134
GenBank mRNA AK001188.1 - 13 - 14 1515 134
AK001726.1 - 9 - 10 1028 134
AK291432.1 - 15 - 16 1849 134
AY267462.1 - 15 - 16 2051 134
BC010218.1 - 11 - 12 1220 134
BC040376.1 - 15 - 16 1773 134
BC047880.1 - 14 - 15 1567 134
BC137438.1 - 15 - 16 2010 134
D28500.1 - 15 - 16 1881 134

View Full Product Details