The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018060.3 | NP_060530.3 | 3 - 4 | 649 | 133 |
GenBank mRNA | AK001188.1 | - | 1 - 2 | 124 | 133 |
AY267462.1 | - | 3 - 4 | 660 | 133 | |
AY952883.1 | - | 3 - 4 | 562 | 133 | |
BC040376.1 | - | 3 - 4 | 382 | 133 | |
BC047880.1 | - | 2 - 3 | 176 | 133 | |
BC137438.1 | - | 3 - 4 | 619 | 133 | |
D28500.1 | - | 3 - 4 | 490 | 133 |