The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016027.2 | NP_057111.1 | 3 - 4 | 502 | 144 |
GenBank mRNA | AF151841.1 | - | 3 - 4 | 445 | 144 |
AK222699.1 | - | 3 - 4 | 455 | 144 | |
AK290201.1 | - | 3 - 4 | 464 | 144 | |
BC000878.1 | - | 3 - 4 | 491 | 144 | |
BC008505.1 | - | 3 - 4 | 491 | 144 |