The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004818.2 | NP_004809.2 | 13 - 14 | 1897 | 105 |
GenBank mRNA | AF026402.1 | - | 13 - 14 | 1845 | 105 |
AK128428.1 | - | 13 - 14 | 1826 | 105 | |
AK292921.1 | - | 13 - 14 | 1886 | 105 | |
AK312379.1 | - | 13 - 14 | 1866 | 105 | |
BC002366.2 | - | 13 - 14 | 1856 | 105 |