The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004818.2 | NP_004809.2 | 15 - 16 | 2152 | 76 |
GenBank mRNA | AF026402.1 | - | 15 - 16 | 2100 | 76 |
AK128428.1 | - | 15 - 16 | 2081 | 76 | |
AK292921.1 | - | 15 - 16 | 2141 | 76 | |
AK312379.1 | - | 15 - 16 | 2121 | 76 | |
BC002366.2 | - | 15 - 16 | 2111 | 76 |