Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COBL
Entrez Gene ID
23242
Gene Name
cordon-bleu WH2 repeat protein
Gene Aliases
-
Chromosome Location
Chr.7: 51016212 - 51316818 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287436.1 NP_001274365.1 1 - 2 230 76
NM_001287438.1 NP_001274367.1 1 - 2 230 76
NM_015198.3 NP_056013.2 1 - 2 230 76
XM_005271750.1 XP_005271807.1 1 - 2 221 76
XM_005271751.1 XP_005271808.1 1 - 2 221 76
XM_005271756.1 XP_005271813.1 1 - 2 221 76
XM_011515234.1 XP_011513536.1 1 - 2 221 76
XM_011515235.1 XP_011513537.1 1 - 2 221 76
XM_011515236.1 XP_011513538.1 1 - 2 221 76
XM_011515237.1 XP_011513539.1 1 - 2 221 76
XM_011515238.1 XP_011513540.1 1 - 2 221 76
XM_011515239.1 XP_011513541.1 1 - 2 221 76
XM_011515240.1 XP_011513542.1 1 - 2 221 76
XM_017011898.1 XP_016867387.1 1 - 2 221 76
XM_017011899.1 XP_016867388.1 1 - 2 221 76
XM_017011900.1 XP_016867389.1 1 - 2 221 76
XM_017011901.1 XP_016867390.1 1 - 2 221 76
GenBank mRNA AB014533.1 - 1 - 2 211 76
AK309254.1 - 1 - 2 221 76
BC094695.1 - 1 - 2 176 76
BC111496.1 - 1 - 2 230 76
BC136441.1 - 1 - 2 145 76
BC144099.1 - 1 - 2 145 76
BC150263.1 - 1 - 2 211 76

View Full Product Details