The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168340.1 | NP_001161812.1 | 5 - 6 | 652 | 119 |
NM_001168341.1 | NP_001161813.1 | 3 - 4 | 445 | 119 | |
NM_001168342.1 | NP_001161814.1 | 3 - 4 | 527 | 119 | |
NM_152574.2 | NP_689787.2 | 5 - 6 | 652 | 119 | |
XM_011517732.2 | XP_011516034.1 | 4 - 5 | 449 | 119 | |
XM_017014310.1 | XP_016869799.1 | 7 - 8 | 1015 | 119 | |
XM_017014311.1 | XP_016869800.1 | 6 - 7 | 947 | 119 | |
XM_017014312.1 | XP_016869801.1 | 4 - 5 | 517 | 119 | |
GenBank mRNA | AK091187.1 | - | 5 - 6 | 538 | 119 |
AK093459.1 | - | 5 - 6 | 719 | 119 | |
AK298581.1 | - | 3 - 4 | 300 | 119 | |
AK298998.1 | - | 5 - 6 | 624 | 119 | |
AK301185.1 | - | 5 - 6 | 503 | 119 | |
AK301427.1 | - | 3 - 4 | 443 | 119 | |
AK310271.1 | - | 6 - 7 | 510 | 119 | |
AK310482.1 | - | 5 - 6 | 652 | 119 | |
BC042417.1 | - | 4 - 5 | 519 | 119 |