The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301088.1 | NP_001288017.1 | 12 - 13 | 1877 | 147 |
NM_019055.5 | NP_061928.4 | 12 - 13 | 2083 | 147 | |
XM_006718861.1 | XP_006718924.1 | 12 - 13 | 2016 | 147 | |
XM_011542875.1 | XP_011541177.1 | 6 - 7 | 845 | 147 | |
GenBank mRNA | AF361473.1 | - | 12 - 13 | 2012 | 147 |
AK027852.1 | - | 12 - 13 | 147 | ||
AK074163.1 | - | 11 - 12 | 1613 | 147 | |
AK289769.1 | - | 12 - 13 | 1982 | 147 | |
AK302628.1 | - | 12 - 13 | 1747 | 147 | |
AY358083.1 | - | 12 - 13 | 1934 | 147 | |
BC014995.1 | - | 11 - 12 | 1695 | 147 | |
BC039602.1 | - | 12 - 13 | 1971 | 147 | |
BC064643.1 | - | 2 - 3 | 230 | 147 | |
BC111562.1 | - | 12 - 13 | 1977 | 147 | |
BC111748.1 | - | 12 - 13 | 1977 | 147 |