The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165979.2 | NP_001159451.1 | 14 - 15 | 3260 | 130 |
NM_001288989.1 | NP_001275918.1 | 15 - 16 | 4701 | 130 | |
NM_016341.3 | NP_057425.3 | 15 - 16 | 4749 | 130 | |
XM_006717885.3 | XP_006717948.1 | 15 - 16 | 4697 | 130 | |
XM_006717888.3 | XP_006717951.1 | 15 - 16 | 4697 | 130 | |
XM_006717889.3 | XP_006717952.1 | 15 - 16 | 4649 | 130 | |
XM_006717890.2 | XP_006717953.1 | 14 - 15 | 3213 | 130 | |
XM_011539849.2 | XP_011538151.1 | 15 - 16 | 5854 | 130 | |
XM_011539850.2 | XP_011538152.1 | 14 - 15 | 4342 | 130 | |
XM_011539851.2 | XP_011538153.1 | 15 - 16 | 4699 | 130 | |
XM_011539852.2 | XP_011538154.1 | 15 - 16 | 4699 | 130 | |
XM_017016310.1 | XP_016871799.1 | 16 - 17 | 4744 | 130 | |
XM_017016311.1 | XP_016871800.1 | 15 - 16 | 4697 | 130 | |
XM_017016312.1 | XP_016871801.1 | 14 - 15 | 3165 | 130 | |
GenBank mRNA | AB040949.2 | - | 15 - 16 | 4749 | 130 |
AF117948.1 | - | 6 - 7 | 824 | 130 | |
AF170071.1 | - | 14 - 15 | 3191 | 130 | |
AF190642.2 | - | 14 - 15 | 4350 | 130 | |
AK289852.1 | - | 14 - 15 | 3213 | 130 | |
BC140705.1 | - | 14 - 15 | 4175 | 130 | |
BC144286.1 | - | 14 - 15 | 4127 | 130 | |
BC151854.1 | - | 15 - 16 | 4749 | 130 |