The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165979.2 | NP_001159451.1 | 17 - 18 | 3651 | 151 |
NM_001288989.1 | NP_001275918.1 | 18 - 19 | 5092 | 151 | |
NM_016341.3 | NP_057425.3 | 18 - 19 | 5140 | 151 | |
XM_006717885.3 | XP_006717948.1 | 18 - 19 | 5088 | 151 | |
XM_006717888.3 | XP_006717951.1 | 18 - 19 | 5088 | 151 | |
XM_006717889.3 | XP_006717952.1 | 18 - 19 | 5040 | 151 | |
XM_006717890.2 | XP_006717953.1 | 17 - 18 | 3604 | 151 | |
XM_011539849.2 | XP_011538151.1 | 18 - 19 | 6245 | 151 | |
XM_011539850.2 | XP_011538152.1 | 17 - 18 | 4733 | 151 | |
XM_011539851.2 | XP_011538153.1 | 18 - 19 | 5090 | 151 | |
XM_011539852.2 | XP_011538154.1 | 18 - 19 | 5090 | 151 | |
XM_017016310.1 | XP_016871799.1 | 19 - 20 | 5135 | 151 | |
XM_017016311.1 | XP_016871800.1 | 18 - 19 | 5088 | 151 | |
XM_017016312.1 | XP_016871801.1 | 17 - 18 | 3556 | 151 | |
GenBank mRNA | AB040949.2 | - | 18 - 19 | 5140 | 151 |
AF117948.1 | - | 9 - 10 | 1215 | 151 | |
AF170071.1 | - | 17 - 18 | 3582 | 151 | |
AF190642.2 | - | 17 - 18 | 4741 | 151 | |
AK289852.1 | - | 17 - 18 | 3604 | 151 | |
BC140705.1 | - | 17 - 18 | 4566 | 151 | |
BC144286.1 | - | 17 - 18 | 4518 | 151 | |
BC151854.1 | - | 18 - 19 | 5140 | 151 |