The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165979.2 | NP_001159451.1 | 3 - 4 | 956 | 64 |
NM_001288989.1 | NP_001275918.1 | 4 - 5 | 2445 | 64 | |
NM_016341.3 | NP_057425.3 | 4 - 5 | 2445 | 64 | |
XM_006717885.3 | XP_006717948.1 | 4 - 5 | 2393 | 64 | |
XM_006717888.3 | XP_006717951.1 | 4 - 5 | 2393 | 64 | |
XM_006717889.3 | XP_006717952.1 | 4 - 5 | 2393 | 64 | |
XM_006717890.2 | XP_006717953.1 | 3 - 4 | 909 | 64 | |
XM_011539849.2 | XP_011538151.1 | 4 - 5 | 3550 | 64 | |
XM_011539850.2 | XP_011538152.1 | 3 - 4 | 2038 | 64 | |
XM_011539851.2 | XP_011538153.1 | 4 - 5 | 2395 | 64 | |
XM_011539852.2 | XP_011538154.1 | 4 - 5 | 2395 | 64 | |
XM_017016310.1 | XP_016871799.1 | 5 - 6 | 2440 | 64 | |
XM_017016311.1 | XP_016871800.1 | 4 - 5 | 2393 | 64 | |
XM_017016312.1 | XP_016871801.1 | 3 - 4 | 909 | 64 | |
GenBank mRNA | AB040949.2 | - | 4 - 5 | 2445 | 64 |
AF170071.1 | - | 3 - 4 | 887 | 64 | |
AF190642.2 | - | 3 - 4 | 2046 | 64 | |
AK289852.1 | - | 3 - 4 | 909 | 64 | |
AK297779.1 | - | 3 - 4 | 696 | 64 | |
BC140705.1 | - | 3 - 4 | 1871 | 64 | |
BC144286.1 | - | 3 - 4 | 1871 | 64 | |
BC151854.1 | - | 4 - 5 | 2445 | 64 |