The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003554.2 | NP_003545.1 | 1 - 1 | 922 | 69 |
GenBank mRNA | BC121105.1 | - | 1 - 1 | 926 | 69 |
BC121106.1 | - | 1 - 1 | 926 | 69 |