The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005502.3 | NP_005493.2 | 3 - 4 | 564 | 61 |
XM_005251773.2 | XP_005251830.1 | 3 - 4 | 473 | 61 | |
XM_005251776.2 | XP_005251833.1 | 2 - 3 | 313 | 61 | |
XM_011518339.2 | XP_011516641.1 | 4 - 5 | 548 | 61 | |
XM_011518340.2 | XP_011516642.1 | 5 - 6 | 763 | 61 | |
XM_011518341.2 | XP_011516643.1 | 4 - 5 | 548 | 61 | |
XM_011518344.2 | XP_011516646.1 | 4 - 5 | 548 | 61 | |
XM_017014378.1 | XP_016869867.1 | 4 - 5 | 746 | 61 | |
XM_017014379.1 | XP_016869868.1 | 4 - 5 | 1154 | 61 | |
XM_017014380.1 | XP_016869869.1 | 4 - 5 | 480 | 61 | |
XM_017014381.1 | XP_016869870.1 | 4 - 5 | 546 | 61 | |
XM_017014382.1 | XP_016869871.1 | 2 - 3 | 1011 | 61 | |
GenBank mRNA | AB037924.1 | - | 2 - 3 | 247 | 61 |
AB055982.1 | - | 2 - 3 | 160 | 61 | |
AB445477.1 | - | 2 - 3 | 160 | 61 | |
AF165281.1 | - | 1 - 2 | 100 | 61 | |
AF258627.1 | - | 3 - 4 | 555 | 61 | |
AF285167.1 | - | 3 - 4 | 450 | 61 | |
AJ012376.1 | - | 1 - 2 | 100 | 61 | |
AK024328.1 | - | 3 - 4 | 473 | 61 | |
BC146856.1 | - | 3 - 4 | 374 | 61 | |
BX955241.1 | - | 3 - 4 | 564 | 61 |