The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032478.3 | NP_115867.2 | 4 - 5 | 817 | 73 |
GenBank mRNA | AK313283.1 | - | 4 - 5 | 632 | 73 |
BC013311.2 | - | 4 - 5 | 616 | 73 | |
BC034460.1 | - | 4 - 5 | 571 | 73 |