Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMNB1
Entrez Gene ID
4001
Gene Name
lamin B1
Gene Aliases
ADLD, LMN, LMN2, LMNB
Chromosome Location
Chr.5: 126776623 - 126837020 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198557.1 NP_001185486.1 7 - 8 1115 101
NM_005573.3 NP_005564.1 7 - 8 2268 101
NR_134488.1 8 - 9 2346 101
GenBank mRNA AK303084.1 - 7 - 8 1115 101
AK312603.1 - 7 - 8 1732 101
BC012295.1 - 7 - 8 1666 101
BC103723.1 - 7 - 8 1763 101
M34458.1 - 7 - 8 1723 101

View Full Product Details