The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171968.1 | NP_001165439.1 | 5 - 6 | 676 | 82 |
NM_021924.4 | NP_068743.2 | 5 - 6 | 676 | 82 | |
NM_031264.3 | NP_112554.2 | 5 - 6 | 676 | 82 | |
XM_006718253.3 | XP_006718316.1 | 5 - 6 | 576 | 82 | |
XM_011520188.2 | XP_011518490.1 | 5 - 6 | 576 | 82 | |
XM_011520189.2 | XP_011518491.1 | 5 - 6 | 576 | 82 | |
XM_011520190.2 | XP_011518492.1 | 5 - 6 | 576 | 82 | |
XM_011520191.2 | XP_011518493.1 | 5 - 6 | 576 | 82 | |
GenBank mRNA | AF258675.1 | - | 6 - 7 | 832 | 82 |
AF258676.1 | - | 6 - 7 | 832 | 82 | |
AF276242.1 | - | 5 - 6 | 511 | 82 | |
AF301909.1 | - | 5 - 6 | 511 | 82 | |
AK000226.1 | - | 5 - 6 | 694 | 82 | |
AK000384.1 | - | 5 - 6 | 581 | 82 | |
AK300991.1 | - | 6 - 7 | 528 | 82 | |
AK308040.1 | - | 5 - 6 | 551 | 82 | |
AK309157.1 | - | 4 - 5 | 471 | 82 | |
AY358368.1 | - | 5 - 6 | 533 | 82 | |
BC038405.1 | - | 5 - 6 | 533 | 82 | |
BC107063.2 | - | 6 - 7 | 82 | ||
DQ491193.1 | - | 5 - 6 | 583 | 82 |