Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD7
Entrez Gene ID
56311
Gene Name
ankyrin repeat domain 7
Gene Aliases
TSA806
Chromosome Location
Chr.7: 118224658 - 118242731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019644.3 NP_062618.2 4 - 5 749 68
GenBank mRNA AK302407.1 - 4 - 5 703 68
BC007020.2 - 2 - 3 283 68
BC032799.1 - 4 - 5 510 68
D78334.1 - 4 - 5 505 68

View Full Product Details