The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270041.1 | NP_001256970.1 | 2 - 3 | 205 | 78 |
NM_001270042.1 | NP_001256971.1 | 2 - 3 | 218 | 78 | |
NM_032372.5 | NP_115748.1 | 3 - 4 | 358 | 78 | |
NR_070308.1 | 4 - 5 | 736 | 78 | ||
NR_070309.1 | 3 - 4 | 518 | 78 | ||
XM_011540267.2 | XP_011538569.1 | 2 - 3 | 177 | 78 | |
XM_011540268.1 | XP_011538570.1 | 2 - 3 | 167 | 78 | |
XM_011540270.2 | XP_011538572.1 | 2 - 3 | 178 | 78 | |
GenBank mRNA | AK093378.1 | - | 3 - 4 | 326 | 78 |
BC007374.2 | - | 3 - 4 | 504 | 78 | |
BC018606.2 | - | 4 - 5 | 736 | 78 | |
BQ229460.1 | - | 2 - 3 | 229 | 78 | |
DA590784.1 | - | 2 - 3 | 205 | 78 |