Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THNSL1
Entrez Gene ID
79896
Gene Name
threonine synthase like 1
Gene Aliases
TSH1
Chromosome Location
Chr.10: 24952319 - 25026664 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005252597.2 XP_005252654.1 1 - 2 197 90

View Full Product Details