The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077365.1 | NP_001070833.1 | 4 - 5 | 479 | 111 |
NM_001077366.1 | NP_001070834.1 | 3 - 4 | 327 | 111 | |
NM_001136113.1 | NP_001129585.1 | 4 - 5 | 719 | 111 | |
NM_007171.3 | NP_009102.3 | 4 - 5 | 479 | 111 | |
XM_005272156.1 | XP_005272213.1 | 4 - 5 | 605 | 111 | |
XM_005272158.1 | XP_005272215.1 | 3 - 4 | 334 | 111 | |
XM_005272162.2 | XP_005272219.1 | 4 - 5 | 453 | 111 | |
XM_017014203.1 | XP_016869692.1 | 4 - 5 | 927 | 111 | |
XM_017014204.1 | XP_016869693.1 | 3 - 4 | 262 | 111 | |
XM_017014205.1 | XP_016869694.1 | 4 - 5 | 401 | 111 | |
GenBank mRNA | AB209430.1 | - | 1 - 2 | 1451 | 293 |
AF095136.1 | - | 4 - 5 | 456 | 111 | |
AK074874.1 | - | 4 - 5 | 437 | 111 | |
AK074888.1 | - | 3 - 4 | 311 | 111 | |
AK301486.1 | - | 4 - 5 | 461 | 111 | |
AK307876.1 | - | 4 - 5 | 437 | 111 | |
AK315218.1 | - | 4 - 5 | 414 | 111 | |
BC022877.2 | - | 4 - 5 | 373 | 111 | |
BC065268.1 | - | 4 - 5 | 664 | 111 |