Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POMT1
Entrez Gene ID
10585
Gene Name
protein O-mannosyltransferase 1
Gene Aliases
LGMD2K, MDDGA1, MDDGB1, MDDGC1, RT
Chromosome Location
Chr.9: 131502895 - 131523806 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077365.1 NP_001070833.1 9 - 10 1059 126
NM_001077366.1 NP_001070834.1 8 - 9 907 126
NM_001136113.1 NP_001129585.1 9 - 10 1299 126
NM_001136114.1 NP_001129586.1 7 - 8 800 126
NM_007171.3 NP_009102.3 9 - 10 1125 126
XM_005272156.1 XP_005272213.1 9 - 10 1251 126
XM_005272158.1 XP_005272215.1 8 - 9 980 126
XM_005272159.1 XP_005272216.1 7 - 8 870 126
XM_005272162.2 XP_005272219.1 8 - 9 1296 126
XM_006716932.1 XP_006716995.1 8 - 9 903 126
XM_011518140.1 XP_011516442.1 8 - 9 978 126
XM_011518141.1 XP_011516443.1 8 - 9 912 126
XM_011518142.1 XP_011516444.1 7 - 8 780 126
XM_011518143.1 XP_011516445.1 7 - 8 805 126
XM_011518145.1 XP_011516447.1 6 - 7 646 126
XM_017014203.1 XP_016869692.1 9 - 10 1573 126
XM_017014204.1 XP_016869693.1 8 - 9 842 126
XM_017014205.1 XP_016869694.1 9 - 10 1047 126
GenBank mRNA AB209430.1 - 4 - 5 2182 126
AF095136.1 - 9 - 10 1102 126
AK074874.1 - 9 - 10 1017 126
AK074888.1 - 8 - 9 891 126
AK293529.1 - 7 - 8 712 126
AK295458.1 - 3 - 4 338 126
AK295561.1 - 7 - 8 804 126
AK299646.1 - 6 - 7 611 126
AK300393.1 - 8 - 9 927 126
AK300511.1 - 1 - 2 186 126
AK303254.1 - 5 - 6 975 126
AK315218.1 - 8 - 9 1257 126
BC022877.2 - 9 - 10 953 126
BC065268.1 - 9 - 10 1244 126

View Full Product Details