Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMX1B
Entrez Gene ID
4010
Gene Name
LIM homeobox transcription factor 1 beta
Gene Aliases
LMX1.2, NPS1
Chromosome Location
Chr.9: 126614443 - 126701032 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174146.1 NP_001167617.1 6 - 7 106
NM_001174147.1 NP_001167618.1 6 - 7 895 73
NM_002316.3 NP_002307.2 6 - 7 895 73
GenBank mRNA AF057135.1 - 6 - 7 819 73
BC069588.1 - 6 - 7 814 73
BC069601.1 - 6 - 7 819 73
BC112120.1 - 6 - 7 869 73
BC113491.1 - 6 - 7 869 73
BC143801.1 - 6 - 7 106
KU178166.1 - 6 - 7 819 73
KU178167.1 - 6 - 7 819 73

View Full Product Details