The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003841.3 | NP_003832.2 | 3 - 4 | 582 | 68 |
GenBank mRNA | AF012536.1 | - | 3 - 4 | 466 | 68 |
AF012629.1 | - | 3 - 4 | 274 | 68 | |
AF014794.1 | - | 3 - 4 | 423 | 68 | |
AF016267.1 | - | 3 - 4 | 461 | 68 | |
AF020502.1 | - | 3 - 4 | 394 | 68 | |
AF033854.1 | - | 3 - 4 | 450 | 68 | |
AK298664.1 | - | 2 - 3 | 369 | 68 | |
AK313213.1 | - | 3 - 4 | 476 | 68 | |
AY358281.1 | - | 3 - 4 | 466 | 68 | |
BC017852.1 | - | 3 - 4 | 475 | 68 | |
BC125041.1 | - | 3 - 4 | 395 | 68 | |
BC125042.1 | - | 3 - 4 | 395 | 68 | |
CR541857.1 | - | 3 - 4 | 274 | 68 |