Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TBATA
- Entrez Gene ID
-
219793
- Gene Name
- thymus, brain and testes associated
- Gene Aliases
-
C10orf27, SPATIAL
- Chromosome Location
-
Chr.10: 70771237 - 70785419 on Build GRCh38
View Full Product Details