The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017629.3 | NP_060099.2 | 11 - 12 | 1575 | 82 |
XM_005270578.3 | XP_005270635.1 | 11 - 12 | 1460 | 82 | |
XM_005270579.3 | XP_005270636.1 | 10 - 11 | 1268 | 82 | |
XM_011540883.2 | XP_011539185.1 | 8 - 9 | 1064 | 82 | |
XM_011540884.2 | XP_011539186.1 | 8 - 9 | 927 | 82 | |
GenBank mRNA | AB046787.1 | - | 11 - 12 | 1575 | 82 |
AK000040.1 | - | 1 - 2 | 64 | 82 | |
AL831908.1 | - | 10 - 11 | 1362 | 82 | |
BC152450.1 | - | 11 - 12 | 1575 | 82 |